Thermoelectric flanging tools, BOLA
Toimittaja: Bohlender
F709-50 F709-52 F709-56 F701-12 F709-54 F701-14
BOHLF709-50EA 117 EUR
BOHLF709-50 BOHLF709-52 BOHLF709-56 BOHLF701-12 BOHLF709-54 BOHLF701-14
Thermoelectric flanging tools, BOLA
Työkalusarjat
Thermoelectric flanging tools are used for making flanges at the ends of plastic tubing such as PTFE, PFA and FEP tubings.
Toimitustiedot: Supplied with basic flanging tool (230 V), exchangeable flanging tip for Int.Ø 0,8 or 1,6 mm and tubing holder for tubing Ext.Ø 1,6 and 3,2 mm.