Tubing connectors, with nut
Toimittaja: Bohlender
D586-18 D585-18 D585-14 D585-25 D586-14
BOHLD586-18EA 5.7 EUR
BOHLD586-18 BOHLD585-18 BOHLD585-14 BOHLD585-25 BOHLD586-14
Tubing connectors, with nut
Kiinnitykset ja liittimet
These connectors are specially designed for a higher flow rate. Connectors made of PP, with elastic sealing lip, FPM o-ring and nut made of PBTP.
  • Available as straight or bent type
  • Universal chemical resistance
  • Maximum temperature up to 250 °C
  • Supplied with matching PBTP cap