Tube connectors, BOLA
Toimittaja: Bohlender
D503-12 D504-04 D504-12 D505-08 D505-12 D526-06 D503-04 D503-08 D526-10 D526-14 D505-06 D503-02 D503-06 D503-18 D504-02 D504-18 D505-02 D505-18 D526-02 D526-04 D526-12 D526-18 D526-20 D526-22 D648-02 D504-08 D505-04 D648-08 D503-14 D505-16 D505-14 D504-16 D503-16 D648-14 D504-14 D526-38 D526-36 D526-34 D526-32 D526-30 D526-28 D526-26
BOHLD503-12EA 44.1 EUR
BOHLD503-12 BOHLD504-04 BOHLD504-12 BOHLD505-08 BOHLD505-12 BOHLD526-06 BOHLD503-04 BOHLD503-08 BOHLD526-10 BOHLD526-14 BOHLD505-06 BOHLD503-02 BOHLD503-06 BOHLD503-18 BOHLD504-02 BOHLD504-18 BOHLD505-02 BOHLD505-18 BOHLD526-02 BOHLD526-04 BOHLD526-12 BOHLD526-18 BOHLD526-20 BOHLD526-22 BOHLD648-02 BOHLD504-08 BOHLD505-04 BOHLD648-08 BOHLD503-14 BOHLD505-16 BOHLD505-14 BOHLD504-16 BOHLD503-16 BOHLD648-14 BOHLD504-14 BOHLD526-38 BOHLD526-36 BOHLD526-34 BOHLD526-32 BOHLD526-30 BOHLD526-28 BOHLD526-26
Tube connectors, BOLA
Kiinnitykset ja liittimet
Tube fitting made of PTFE with nuts made of glass-fibre reinforced PTFE.
  • Universal chemical resistance
  • Maximum temperature up to 250 °C
  • Vacuum: suitable
  • Connects tubing or tube with different outer diameters
  • Features two connections (elbow) and three connections (T- piece)