Stirrer shafts, gassing, with blade, BOLA
Toimittaja: Bohlender
C488-08 C488-14 C488-20
BOHLC488-08EA 778 EUR
BOHLC488-08 BOHLC488-14 BOHLC488-20
Stirrer shafts, gassing, with blade, BOLA
Sekoittajat Sekoitussauvat
Stirrer shafts made of PTFE-jacketed stainless steel and the propeller with four blades completely made of PTFE.
  • Chemical resistant
  • Temperature resistance from –200 to +250 °C
  • Rotation speed depends on the fluid level and the immersion depth
  • Creates strong radial flow, suitable for gassing of liquids
  • With chucking diameter of 8 mm
Clockwise rotation of the shaft produces a vacuum behind the stirrer blades. By this vacuum, the gas is transported from the gas compartment through the hollow shaft and into the product.