Stirrer shafts, solo, BOLA
Toimittaja: Bohlender
C472-08 C474-08 C472-20 C474-30 C474-40 C474-20 C474-34 C476-60 C476-40
BOHLC472-08EA 103 EUR
BOHLC472-08 BOHLC474-08 BOHLC472-20 BOHLC474-30 BOHLC474-40 BOHLC474-20 BOHLC474-34 BOHLC476-60 BOHLC476-40
Stirrer shafts, solo, BOLA
Sekoittajat Sekoitussauvat
Stirrer shafts made of PTFE-jacketed stainless steel with fused lower end.
  • Usable as stirring rod for manual stirring
  • Chemical resistant
  • Temperature resistance from –200 to +250 °C