Stirrer shafts, KPG®, BOLA
Toimittaja: Bohlender
C387-05 C387-07 C387-09 C387-15 C387-13 C387-11
BOHLC387-05EA 347 EUR
BOHLC387-05 BOHLC387-07 BOHLC387-09 BOHLC387-15 BOHLC387-13 BOHLC387-11
Stirrer shafts, KPG®, BOLA
Sekoittajat Sekoitussauvat
KPG stirrer shafts with double pivot made of ground and polished borosilicate glass, tiltable moon-shaped stirrer blade with double-sided groove completely made of PTFE.
  • Chemical resistant
  • Temperature resistance from –200 to +250 °C
  • Creates tangential flow with little turbulence
  • With chucking diameter of 8 mm
Tilting half-moon blade is ideal for stirring in round-bottom flasks with ground joint neck.