Stirrer shafts, KPG®, GT, BOLA
Toimittaja: Bohlender
C375-02 C375-04 C375-06 C375-08 C375-10
BOHLC375-02EA 172 EUR
BOHLC375-02 BOHLC375-04 BOHLC375-06 BOHLC375-08 BOHLC375-10
Stirrer shafts, KPG®, GT, BOLA
Sekoittajat Sekoitussauvat
KPG stirrer shafts made of borosilicate glass, tiltable moon-shaped stirrer blade with angular groove and clamping bolts completely made of PTFE.
  • Chemical resistant
  • Temperature resistance from –200 to +250 °C
  • Creates tangential flow with little turbulence
  • With chucking diameter of 8 mm
Tilting half-moon blade is ideal for stirring in round-bottom flasks with ground joint neck.