Bearing neck for tandem magnetic stirring bar, BOLA
Toimittaja: Bohlender
C364-16 C364-08
BOHLC364-16EA 33.5 EUR
BOHLC364-16 BOHLC364-08
Bearing neck for tandem magnetic stirring bar, BOLA
Sekoittajat Sekoitustyökalut
Bearing necks for receiving tandem magnetic stirring bar. The centre portion will fix to the vessel by means of glue such as silicone hardened in water.
  • Chemical resistance
  • Made of very hard PTFE/PEEK compound
Temperature resistance: −100 to +240 °C.