Dekantteri, matala malli
Toimittaja: Bohlender
A137-09 A137-07 A137-05 A137-03 A137-02 A137-01
BOHLA137-09EA 406 EUR
BOHLA137-09 BOHLA137-07 BOHLA137-05 BOHLA137-03 BOHLA137-02 BOHLA137-01
Dekantteri, matala malli
Dekantterit
PFA, translucent, with spout.
  • Space-saving stackable design
  • High thermal resistance from -200 to +250 °C
  • Autoclavable, easy to clean
Embossed graduations are easy to read and resistant to wear from repeated use.
Sertifikaatit: FDA conform.