Dekantteri, matala malli
Toimittaja: Bohlender
A136-14 A136-15 A136-06 A136-08 A136-04 A136-13 A136-11 A136-09 A136-05 A136-03 A136-02 A136-07
BOHLA136-14EA 487 EUR
BOHLA136-14 BOHLA136-15 BOHLA136-06 BOHLA136-08 BOHLA136-04 BOHLA136-13 BOHLA136-11 BOHLA136-09 BOHLA136-05 BOHLA136-03 BOHLA136-02 BOHLA136-07
Dekantteri, matala malli
Dekantterit
PTFE, with spout.
  • Excellent chemical resistance
  • Temperature resistant from –200 to +250 °C
  • Flat bottom for good heat transfer, for example when used on regulated hotplates
Thick walled, with reinforced lip and exceptionally smooth inner walls to prevent contamination. Non stick. Low gas permeability.