Overhead stirrer tools, turbine stirrers
Toimittaja: BOCHEM
9025 9026
BOCH9025EA 155 EUR
BOCH9025 BOCH9026
Overhead stirrer tools, turbine stirrers
Sekoittajat Sekoitustyökalut
Propeller stirrers made of stainless steel, with ring.
  • Ideal for stirring liquids at middle speed range
  • Suitable for stirring low and medium viscous media
  • Creates axial flow
  • Ideal for homogenising and suspensioning