Dekantterit, matala malli
Toimittaja: BOCHEM
8505 8506 8531 8532 8514 8526 8516 8520 8521 8522 8523 8525 8530 8533 8534 8535 8536 8524 8513 8515 8502 8501 8503 8504 8510 8511 8512 8500
BOCH8505EA 400 EUR
BOCH8505 BOCH8506 BOCH8531 BOCH8532 BOCH8514 BOCH8526 BOCH8516 BOCH8520 BOCH8521 BOCH8522 BOCH8523 BOCH8525 BOCH8530 BOCH8533 BOCH8534 BOCH8535 BOCH8536 BOCH8524 BOCH8513 BOCH8515 BOCH8502 BOCH8501 BOCH8503 BOCH8504 BOCH8510 BOCH8511 BOCH8512 BOCH8500
Dekantterit, matala malli
Dekantterit
Stainless steel, 18/10.
  • Good chemical resistance
  • Excellent resistance to heat
  • Smooth surfaces for fast, easy cleaning
Ideal for corrosive materials. Robust and durable.