Bowls
Toimittaja: BOCHEM
8445 8446 8447 8448
216-0020EA 213 EUR
216-0020 216-0010 216-0021 BOCH8448
Bowls
Haihdutusmaljat/Tarjottimet/Altaat Laboratorioalustat/-astiat
Made of stainless steel with two handles.
  • Very good chemical resistance
  • Very good thermal resistance
  • Smooth surfaces for easy cleaning
Multipurpose bowls ideal for a wide variety of uses.
Order Now
TEKNISET TIEDOT
Environmentally Preferable