Test tube stands, round
Toimittaja: BOCHEM
8230 8231 8233
BOCH8230EA 104 EUR
BOCH8230 BOCH8231 BOCH8233
Test tube stands, round
Koeputkitelineet Tube Racks
Rack, made of stainless steel 18/10.
  • With handle
  • Very good tempearture resistance
  • Good chemical resistance