Pharmacist spoons
Toimittaja: BOCHEM
3360 3370 3380 3385
BOCH3360EA 7.4 EUR
BOCH3360 231-1281 231-1282 BOCH3385
Pharmacist spoons
Lusikat
Stainless steel 18/10, anti-magnetic. Multipurpose spoons for a wide range of uses.
  • Narrow shape with extra long, flat handle
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
  • Quick and easy to clean