Spatulas, round, bent
Toimittaja: BOCHEM
3200 3202 3203 3204 32005 3201 32025
231-1045EA 4.3 EUR
231-1045 231-1047 BOCH3203 BOCH3204 BOCH32005 BOCH3201 BOCH32025
Spatulas, round, bent
Lastat
Stainless steel ¹⁸/₁₀, anti-magnetic. These spatulas with one rounded end can be used for a variety of applications.
  • Excellent heat resistance, autoclavable
  • Good resistance to chemicals
  • Easy and quick cleaning