Flask tongs, stainless steel
Toimittaja: BOCHEM
2180 2179 2183 2182 2181
BOCH2180EA 31.1 EUR
BOCH2180 232-4153 BOCH2183 BOCH2182 BOCH2181
Flask tongs, stainless steel
Pihdit
Flask tongs made of electropolished stainless steel with polyamide coated endings.