Verkkokorit kuljetusta varten
Toimittaja: BOCHEM
17300 17301 17302 17305 17306 17307 17310 17311 17312
BOCH17300EA 666 EUR
BOCH17300 BOCH17301 BOCH17302 BOCH17305 BOCH17306 BOCH17307 BOCH17310 BOCH17311 BOCH17312
Verkkokorit kuljetusta varten
Korit Korit, lanka
Stainless steel 18/10.
  • Three different mesh dimensions
  • Stackable
  • Excellent thermal and chemical resistance
These rectangular baskets are designed for transporting, storing, cleaning and sterilising laboratory utensils.