Support clamps
Toimittaja: BOCHEM
17210 17211 17212 17213 17214 17215 17216
BOCH17210EA 1510 EUR
BOCH17210 BOCH17211 BOCH17212 BOCH17213 BOCH17214 BOCH17215 BOCH17216
Support clamps
Kourat Yleiskäyttöiset puristimet
Support clamps made of stainless steel (18/19) and PTFE for either Schott laboratory flat flanges (PF) (optimal flange angle 45°) or Schott "Kugel-Flanges" (ball and socket system) (KF).
  • For reactors and other vessels
  • User friendly thanks to joint screws
  • With two supporting devices for 26,9 mm tubes