PF-laboratory flange and KF-support clamps
Toimittaja: BOCHEM
17200 17201 17202 17203 17204 17207 17205 17206
BOCH17200EA 1440 EUR
BOCH17200 BOCH17201 BOCH17202 BOCH17203 BOCH17204 BOCH17207 BOCH17205 BOCH17206
PF-laboratory flange and KF-support clamps
Kourat Yleiskäyttöiset puristimet
Support clamps made of stainless steel (18/19) and PTFE for either Schott laboratory flat flanges (PF) (optimal flange angle 45°) or Schott 'Kugel-Flanges' (ball and socket system) (KF).
  • For reactors and other vessels
  • User friendly through joint screws
  • With two support bars 15×200 mm