Laboratory tables, without ground plate
Toimittaja: BOCHEM
11332 11333 11334 11302 11301 11303 11304 11300 11532 11533 11534 11502 11501 11503 11504 11500
BOCH11332EA 8380 EUR
BOCH11332 BOCH11333 BOCH11334 BOCH11302 BOCH11301 BOCH11303 BOCH11304 BOCH11300 BOCH11532 BOCH11533 BOCH11534 BOCH11502 BOCH11501 BOCH11503 BOCH11504 BOCH11500
Laboratory tables, without ground plate
Kalusteet Pöydät
These laboratory tables are made up of 18/10 stainless steel and height adjustable.
  • Drawer face with user friendly handle bar
  • Noise protection
  • Worktops underlaid with soundproofing acoustic wood
Working plate with matt finishing and corners are continuously welded and polished.
Drawer with guard rails made of stainless steel to mount GN-drawer. Drawers can be taken off for cleaning, but are secured against drawing out.