Anti-CA19-9 Mouse Monoclonal Antibody [clone: 121SLE]
Toimittaja: Biotium
BNUM0025-50
BNUM0025-50EA 417 EUR
BNUM0025-50
Anti-CA19-9 Mouse Monoclonal Antibody [clone: 121SLE]
Vasta-aineet
CA19-9, a carbohydrate epitope expressed on a high MW (>400 kDa) mucin glycoprotein, is a sialyl Lewisa structure which is synthesized from type 1 blood group precursor chains and is present in individuals expressing the Lewisa and/or Lewisb blood group antigens. In normal tissues, sialyl Lewisa antigen is present in ductal epithelium of the breast, kidney, salivary gland, and sweat glands. Its expression is greatly enhanced in serum as well as in the majority of tumor cells in gastrointestinal (GI) carcinomas, including adenocarcinomas of the stomach, intestine, and pancreas.More Product Information