Anti-KRT19 Mouse Monoclonal Antibody (Purified) [clone: KRT19/799]
Toimittaja: Biotium
BNUB0799-500 BNUB0799-100
BNUB0799-500EA 511 EUR
BNUB0799-500 BNUB0799-100
Anti-KRT19 Mouse Monoclonal Antibody (Purified) [clone: KRT19/799]
Vasta-aineet
Recognizes a protein of 40 kDa, identified as cytokeratin-19 (CK19), which is expressed in sweat gland, mammary gland ductal and secretory cells, bile ducts, gastrointestinal tract, bladder urothelium, oral epithelia, esophagus, and ectocervical epithelium. Anti-CK19 reacts with a wide variety of epithelial malignancies including adenocarcinomas of the colon, stomach, pancreas, biliary tract, liver, and breast. Perhaps the most useful application is the identification of thyroid carcinoma of the papillary type, although 50%-60% of follicular carcinomas are also labeled. Anti-CK19 is a usefulMore Product Information