Anti-CDKN1B Mouse Monoclonal Antibody (Purified) [clone: DCS-72.F6]

Toimittaja: Biotium
BNUB0771-500 BNUB0771-100
BNUB0771-500EA 511 EUR
BNUB0771-500 BNUB0771-100
Anti-CDKN1B Mouse Monoclonal Antibody (Purified) [clone: DCS-72.F6]
Vasta-aineet
Recognizes a 27 kDa protein, identified as the p27Kip1, a cell cycle regulatory mitotic inhibitor. Its epitope spans between aa 83-204 of p27. It is highly specific and shows no cross-reaction with other related mitotic inhibitors. p27Kip1 functions as a negative regulator of G1 progression and has been proposed to function as a possible mediator of TGF- induced G1 arrest. p27Kip1 is a candidate tumor suppressor gene. This MAb co-precipitates cdk4 in complex p27Kip1 and is excellent for staining of formalin-fixed tissues.

Type: Primary
Antigen: CDKN1B
Clonality: Monoclonal
Clone: DCS-72.F6
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR