Anti-CD66 molecule Mouse Monoclonal Antibody (Purified) [clone: CEA31]
Toimittaja: Biotium
BNUB0670-500 BNUB0670-100
BNUB0670-500EA 511 EUR
BNUB0670-500 BNUB0670-100
Anti-CD66 molecule Mouse Monoclonal Antibody (Purified) [clone: CEA31]
Vasta-aineet
This antibody recognizes proteins of 80-200 kDa, identified as different members of CEA family. CEA is synthesized during development in the fetal gut and is re-expressed in increased amounts in intestinal carcinomas and several other tumors. This MAb does not react with nonspecific cross-reacting antigen (NCA) and with human polymorphonuclear leucocytes. It shows no reaction with a variety of normal tissues and is suitable for staining of formalin/paraffin tissues. CEA is not found in benign glands, stroma, or malignant prostatic cells. Antibody to CEA is useful in detecting early foci ofMore Product Information