Anti-NCL Mouse Monoclonal Antibody (Purified) [clone: 364-5]
Toimittaja: Biotium
BNUB0527-500 BNUB0527-100
BNUB0527-500EA 511 EUR
BNUB0527-500 BNUB0527-100
Anti-NCL Mouse Monoclonal Antibody (Purified) [clone: 364-5]
Vasta-aineet
Recognizes a protein of ~76 kDa, which is identified as Nucleolin (NCL). It is the major nucleolar phosphoprotein of growing eukaryotic cells. NCL is located mainly in dense fibrillar regions of the nucleolus. It is found associated with intranucleolar chromatin and pre-ribosomal particles. Human NCL gene consists of 14 exons with 13 introns and spans approximately 11kb. It induces chromatin decondensation by binding to histone H1. It is thought to play a role in pre-rRNA transcription and ribosome assembly.This MAb can be used to stain the nucleoli in cell or tissue preparations and can beMore Product Information