Anti-CD4 Mouse Monoclonal Antibody (Biotin) [clone: EDU-2]
Toimittaja: Biotium
BNCB0345-500 BNCB0345-100
BNCB0345-500EA 560 EUR
BNCB0345-500 BNCB0345-100
Anti-CD4 Mouse Monoclonal Antibody (Biotin) [clone: EDU-2]
Vasta-aineet
Recognizes a protein of 55 kDa, identified as CD4. It is a membrane glycoprotein of T lymphocytes that interacts with major histocompatibility complex class II antigens and is also a receptor for the human immunodeficiency virus. This protein is expressed not only in T lymphocytes, but also in B cells, macrophages, and granulocytes. It is also expressed in specific regions of the brain. The protein functions to initiate or augment the early phase of T-cell activation, and may function as an important mediator of indirect neuronal damage in infectious and immune-mediated diseases of theMore Product Information