Anti-KRT19 Mouse Monoclonal Antibody (Biotin) [clone: A53-B/A2.26]
Toimittaja: Biotium
BNCB0020-500 BNCB0020-100
BNCB0020-500EA 560 EUR
BNCB0020-500 BNCB0020-100
Anti-KRT19 Mouse Monoclonal Antibody (Biotin) [clone: A53-B/A2.26]
Vasta-aineet
This MAb reacts with the rod domain of human cytokeratin-19 (CK19), a polypeptide of 40 kDa. Its epitope maps between amino acid 312-335. CK19 is expressed in sweat gland, mammary gland ductal and secretory cells, bile ducts, gastrointestinal tract, bladder urothelium, oral epithelia, esophagus, and ectocervical epithelium. Anti-CK19 reacts with a wide variety of epithelial malignancies including adenocarcinomas of the colon, stomach, pancreas, biliary tract, liver, and breast. Perhaps the most useful application is the identification of thyroid carcinoma of the papillary type, although 50%-6More Product Information