Anti-HLA-DPB1 & HLA-DRA Mouse Monoclonal Antibody (CF594) [clone: Bra-14]
Toimittaja: Biotium
BNC941129-500 BNC941129-100
BNC941129-500EA 560 EUR
BNC941129-500 BNC941129-100
Anti-HLA-DPB1 & HLA-DRA Mouse Monoclonal Antibody (CF594) [clone: Bra-14]
Vasta-aineet
Reacts with a common epitope of human major histocompatibility (MHC) class II antigens, HLA-DR and DP. Human MHC class II antigens are transmembrane glycoproteins composed of an chain (36 kDa) and a chain (27 kDa). They are expressed primarily on antigen presenting cells such as B lymphocytes, monocytes, macrophages, and thymic epithelial cells and are also present on activated T lymphocytes. Human MHC class II genes are located in the HLA-D region that encodes at least six and ten chain genes. Three loci, DR, DQ and DP, encode the major expressed products of the human class II region. TheMore Product Information