Anti-S100A9 Mouse Monoclonal Antibody (CF594) [clone: S100A9/1075]
Toimittaja: Biotium
BNC941075-500 BNC941075-100
BNC941075-500EA 560 EUR
BNC941075-500 BNC941075-100
Anti-S100A9 Mouse Monoclonal Antibody (CF594) [clone: S100A9/1075]
Vasta-aineet
This MAb stains the cytoplasm of macrophages and histiocytes in hematopoietic organs, Kupffer's cells of the liver and Langerhan's cells of the skin. It also stains the mantle zone B-lymphocytes of the lymph node and spleen, spermatogonia, and chief cells of the stomach. S100A9 is expressed by macrophages in acutely inflamed tissues and in chronic inflammation. It is detected in peripheral blood leukocytes, in neutrophils and granulocytes. It is present at sites of vascular inflammation. S100A9 is also expressed in epithelial cells constitutively or induced during dermatoses. S100A9 is aMore Product Information