Anti-CD66 molecule Mouse Monoclonal Antibody (CF594) [clone: C66/1009]
Toimittaja: Biotium
BNC941009-500 BNC941009-100
BNC941009-500EA 560 EUR
BNC941009-500 BNC941009-100
Anti-CD66 molecule Mouse Monoclonal Antibody (CF594) [clone: C66/1009]
Vasta-aineet
This antibody recognizes proteins of 80-200 kDa, identified as different members of CEA family. CEA is synthesized during development in the fetal gut and is re-expressed in increased amounts in intestinal carcinomas and several other tumors. This MAb does not react with nonspecific cross-reacting antigen (NCA) and with human polymorphonuclear leucocytes. It shows no reaction with a variety of normal tissues and is suitable for staining of formalin/paraffin tissues. CEA is not found in benign glands, stroma, or malignant prostatic cells. Antibody to CEA is useful in detecting early foci ofMore Product Information
Order Now
TEKNISET TIEDOT
Antigeenin symboli CD66 molecule
Antigeenin nimi CD66
Environmentally Preferable
Antigeenin synonyymit CEACAM5|CD66|Biliary Glycoprotein (BGP-1)|Carcinoembryonic Antigen-related Cell Adhesion Molecule 5
Vasta-aineen tyyppi Ensisijainen
Klonaalisuus Monoclonal
Konjugaatio CF® 594
Clone Clone C66/1009
Reaktiivisuus Human
Isäntä Mouse
Geeni ID 709196
Isotyyppi IgG2a kappa
ImmunoKemia Yes
ImmunoFluoresenssi Yes
Pitoisuus 0,1 mg/ml
Säilytyspuskuri PBS/0,1% BSA/0,05% azide