Anti-CD68 Mouse Monoclonal Antibody (CF594) [clone: CD68/G2]
Toimittaja: Biotium
BNC940919-500 BNC940919-100
BNC940919-500EA 560 EUR
BNC940919-500 BNC940919-100
Anti-CD68 Mouse Monoclonal Antibody (CF594) [clone: CD68/G2]
Vasta-aineet
This antibody recognizes a glycoprotein of 110 kDa, which is identified as CD68. It is important for identifying macrophages in tissue sections. It stains macrophages in a wide variety of human tissues, including Kupffer cells and macrophages in the red pulp of the spleen, in lamina propria of the gut, in lung alveoli, and in bone marrow. It reacts with myeloid precursors and peripheral blood granulocytes. It also reacts with plasmacytoid T cells, which are supposed to be of monocyte/macrophage origin. It shows strong granular cytoplasmic staining of chronic and acute myeloid leukemia andMore Product Information