Anti-NKX2-1 Mouse Monoclonal Antibody (CF594) [clone: 8G7G3/1 NX2.1/690]
Toimittaja: Biotium
BNC940907-500 BNC940907-100
BNC940907-500EA 560 EUR
BNC940907-500 BNC940907-100
Anti-NKX2-1 Mouse Monoclonal Antibody (CF594) [clone: 8G7G3/1 NX2.1/690]
Vasta-aineet
Recognizes a protein of 40 kDa, identified as Thyroid transcription factor-1 (TTF-1). TTF-1 is a member of the NKx2 family of homeodomain transcription factors. It is expressed in epithelial cells of the thyroid gland and the lung. Nuclei from liver, stomach, pancreas, small intestine, colon, kidney, breast, skin, testes, pituitary, prostate, and adrenal glands are unreactive. Anti-TTF-1 is useful in differentiating primary adenocarcinoma of the lung from metastatic carcinomas originating in the breast, mediastinal germ cell tumors, and malignant mesothelioma. It can also be used toMore Product Information