Anti-CHGA Mouse Monoclonal Antibody (CF594) [clone: LK2H10]
Toimittaja: Biotium
BNC940641-500 BNC940641-100
BNC940641-500EA 560 EUR
BNC940641-500 BNC940641-100
Anti-CHGA Mouse Monoclonal Antibody (CF594) [clone: LK2H10]
Vasta-aineet
Chromogranin A is present in neuroendocrine cells throughout the body, including the neuroendocrine cells of the large and small intestine, adrenal medulla and pancreatic islets. It is an excellent marker for carcinoid tumors, pheochromocytomas, paragangliomas, and other neuroendocrine tumors. Co-expression of chromogranin A and neuron specific enolase (NSE) is common in neuroendocrine neoplasms. Reportedly, co-expression of certain keratins and chromogranin indicates neuroendocrine lineage. The presence of strong anti-chromogranin staining and absence of anti-keratin staining should raiseMore Product Information