Anti-ESR2 Mouse Monoclonal Antibody (CF594) [clone: ERb455]
Toimittaja: Biotium
BNC940455-500 BNC940455-100
BNC940455-500EA 560 EUR
BNC940455-500 BNC940455-100
Anti-ESR2 Mouse Monoclonal Antibody (CF594) [clone: ERb455]
Vasta-aineet
Estrogen receptors (ER) are members of the steroid/thyroid hormone receptor superfamily of ligand-activated transcription factors. Estrogen receptors, including ER-alpha and ER-beta, contain DNA binding and ligand binding domains and are critically involved in regulating the normal function of reproductive tissues. They are located in the nucleus, though some estrogen receptors associate with the cell surface membrane and can be rapidly activated by exposure of cells to estrogen. ER-alpha and ER-beta are differentially activated by various ligands. Receptor-ligand interactions trigger aMore Product Information