Anti-KRT77 & KRT76 Mouse Monoclonal Antibody (CF594) [clone: AE-1 / AE-3]
Toimittaja: Biotium
BNC940371-500 BNC940371-100
BNC940371-500EA 560 EUR
BNC940371-500 BNC940371-100
Anti-KRT77 & KRT76 Mouse Monoclonal Antibody (CF594) [clone: AE-1 / AE-3]
Vasta-aineet
Twenty human keratins are resolved with two-dimensional gel electrophoresis into acidic (pI 6.0) subfamilies. This antibody cocktail recognizes acidic (Type I or LMW) and basic (Type II or HMW) cytokeratins, which 67 kDa (CK1); 64 kDa (CK3); 59 kDa (CK4); 58 kDa (CK5); 56 kDa (CK6); 52 kDa (CK8); 56.5 kDa (CK10); 50 kDa (CK14); 50 kDa (CK15); 48 kDa (CK16); 40 kDa (CK19). Many studies have shown the usefulness of keratins as markers in cancer research and tumor diagnosis. AE-1/AE-3 is a broad spectrum anti pan-cytokeratin antibody cocktail, which differentiates epithelial tumors from non-More Product Information