Anti-MUC1 Mouse Monoclonal Antibody (CF594) [clone: GP1.4]
Toimittaja: Biotium
BNC940159-500 BNC940159-100
BNC940159-500EA 560 EUR
BNC940159-500 BNC940159-100
Anti-MUC1 Mouse Monoclonal Antibody (CF594) [clone: GP1.4]
Vasta-aineet
In Western blotting, this antibody recognizes proteins in MW range of 265-400 kDa, identified as different glycoforms of EMA. This MAb reacts with the DTRP epitope in the tandem repeats. The α subunit has cell adhesive properties. It can act both as an adhesion and an anti-adhesion protein. EMA may provide a protective layer on epithelial cells against bacterial and enzyme attack. The β subunit contains a C-terminal domain, which is involved in cell signaling, through phosphorylations and protein-protein interactions. In immunohistochemical assays, it superbly stains routine formalin/More Product Information