Anti-HLA-DRB1 Mouse Monoclonal Antibody (CF488A) [clone: LN-3 HLA-DRB/1067]
Toimittaja: Biotium
BNC881208-500 BNC881208-100
BNC881208-500EA 560 EUR
BNC881208-500 BNC881208-100
Anti-HLA-DRB1 Mouse Monoclonal Antibody (CF488A) [clone: LN-3 HLA-DRB/1067]
Vasta-aineet
This MAb reacts with the beta-chain of HLA-DRB1 antigen, a member of MHC class II molecules. It does not cross react with HLA-DP and HLA-DQ. HLA-DR is a heterodimeric cell surface glycoprotein comprised of a 36 kDa alpha (heavy) chain and a 28 kDa beta (light) chain. It is expressed on B-cells, activated T-cells, monocytes/macrophages, dendritic cells and other non-professional APCs. In conjunction with the CD3/TCR complex and CD4 molecules, HLA-DR is critical for efficient peptide presentation to CD4 T cells. It is an excellent histiocytic marker in paraffin sections producing intenseMore Product Information