Anti-CD70 Mouse Monoclonal Antibody (CF488A) [clone: TNFS7/1026]

Toimittaja: Biotium
BNC881026-500 BNC881026-100
BNC881026-500EA 560 EUR
BNC881026-500 BNC881026-100
Anti-CD70 Mouse Monoclonal Antibody (CF488A) [clone: TNFS7/1026]
Vasta-aineet
This antibody recognizes a protein of 30 kDa, identified as CD70. It is a cytokine that belongs to the tumor necrosis factor (TNF) ligand family. This cytokine is a ligand for TNFRSF27/CD27. It is a surface antigen on activated, but not on resting, T- and B-lymphocytes. It induces proliferation of co-stimulated T cells, enhances the generation of cytolytic T cells, and contributes to T cell activation. This cytokine is also reported to play a role in regulating B-cell activation, cytotoxic function of natural killer cells, and immunoglobulin synthesis.

Type: Primary
Antigen: CD70
Clonality:More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR