Anti-NGFR Mouse Monoclonal Antibody (CF488A) [clone: NGFR5 NTR/912]
Toimittaja: Biotium
BNC880914-500 BNC880914-100
BNC880914-500EA 560 EUR
BNC880914-500 BNC880914-100
Anti-NGFR Mouse Monoclonal Antibody (CF488A) [clone: NGFR5 NTR/912]
Vasta-aineet
This antibody recognizes a glycoprotein of 75 kDa, identified as low affinity Nerve Growth Factor (NGF) Receptor (p75NGFR) or Neurotrophin Receptor (p75NTR). NGFR is expressed in various neural crest cells and their tumors such as melanocytes, melanomas, neuroblastomas, pheochromocytomas and neurofibromas. Reportedly, anti-NGFR is a reliable marker for desmoplastic and neurotropic melanomas. NGFR is expressed in mature non-neural cells such as perivascular cells, dental pulp cells, lymphoidal follicular dendritic cells, basal epithelium of oral mucosa and hair follicles, prostate basal cells,More Product Information
Order Now
TEKNISET TIEDOT
Antigeenin symboli NGFR
Environmentally Preferable
Antigeenin nimi Nerve Growth Factor Receptor
Vasta-aineen tyyppi Ensisijainen
Klonaalisuus Monoclonal
Konjugaatio CF® 488A
Clone Clone NGFR5 NTR/912
Reaktiivisuus Primates,Human
Isäntä Mouse
Geeni ID 415768 , 681726
Isotyyppi IgG
ImmunoKemia Yes
ImmunoFluoresenssi Yes
Pitoisuus 0,1 mg/ml
Säilytyspuskuri PBS/0,1% BSA/0,05% azide