Anti-CD63 Mouse Monoclonal Antibody (CF488A) [clone: MX-49.129.5]
Toimittaja: Biotium
BNC880525-500 BNC880525-100
BNC880525-500EA 560 EUR
BNC880525-500 BNC880525-100
Anti-CD63 Mouse Monoclonal Antibody (CF488A) [clone: MX-49.129.5]
Vasta-aineet
This MAb recognizes protein of 26 kDa-60 kDa, which is identified as CD63. Its epitope is different from that of MAb LAMP3/529. The tetraspanins are integral membrane proteins expressed on cell surface and granular membranes of hematopoietic cells and are components of multi-molecular complexes with specific integrins. The tetraspanin CD63 is a lysosomal membrane glycoprotein that translocates to the plasma membrane after platelet activation. CD63 is expressed on activated platelets, monocytes and macrophages, and is weakly expressed on granulocytes, T cell and B cells. It is located on theMore Product Information