Anti-IGHM Mouse Monoclonal Antibody (CF488A) [clone: IM260]
Toimittaja: Biotium
BNC880260-500 BNC880260-100
BNC880260-500EA 560 EUR
BNC880260-500 BNC880260-100
Anti-IGHM Mouse Monoclonal Antibody (CF488A) [clone: IM260]
Vasta-aineet
Recognizes a protein of 75 kDa, identified as mu heavy chain of human immunoglobulins. It does not cross-react with alpha (IgA), gamma (IgG), epsilon (IgE), or delta (IgD), heavy chains, T-cells, monocytes, granulocytes, or erythrocytes. This MAb is useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkin's lymphomas. The most common feature of these malignancies is the restricted expression of a single heavy chain class. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant.
Type: Primary
Antigen: IGHM