Anti-Progesterone Mouse Monoclonal Antibody (CF488A) [clone: 6-5E-3F]
Toimittaja: Biotium
BNC880236-500 BNC880236-100
BNC880236-500EA 560 EUR
BNC880236-500 BNC880236-100
Anti-Progesterone Mouse Monoclonal Antibody (CF488A) [clone: 6-5E-3F]
Vasta-aineet
This MAb is specific for progesterone. It exhibits minimal cross reactivity with related compounds in ELISA. It reacts with Progesterone-11a-HMS-BSA: 100%; 5-beta-Pregnane-3,20-dione: 48%; 5-alpha-Pregnane-3,20-dione: 26.4%; 17-alpha-Hydroxyprogesterone: 2.5%; 20-alpha-Hydroxyprogesterone: 0.04%. Progesterone is a steroid hormone synthesized from the cholesterol derivative, pregnenolone, in the cortex of the adrenal gland. Progesterone is secreted by the corpus luteum and acts to prepare the endometrium for the implantation of a fertilized egg. During pregnancy, it is secreted by theMore Product Information