Anti-CKB Mouse Monoclonal Antibody (CF488A) [clone: 2ba6]
Toimittaja: Biotium
BNC880196-500 BNC880196-100
BNC880196-500EA 560 EUR
BNC880196-500 BNC880196-100
Anti-CKB Mouse Monoclonal Antibody (CF488A) [clone: 2ba6]
Vasta-aineet
Creatine kinases (CK) are a large family of isoenzymes that regulate levels of ATP in subcellular compartments, where they provide ATP at sites of fluctuating energy demand by the transfer of phosphates between creatine and adenine nucleotides. CKs provide the energy of phosphate hydrolysis necessary to drive the normal function of many cellular systems. In cells, the cytosolic CK enzymes consist of two subunits, which can be either B (brain type) or M (muscle type). There are three different isoenzymes: CKMM, CKBB and CKMB. This MAb recognizes the CKBB isoenzyme and does not react with theMore Product Information