Anti-CMV-p65 Mouse Monoclonal Antibody (CF488A) [clone: CMV101]

Toimittaja: Biotium
BNC880101-500 BNC880101-100
BNC880101-500EA 560 EUR
BNC880101-500 BNC880101-100
Anti-CMV-p65 Mouse Monoclonal Antibody (CF488A) [clone: CMV101]
Vasta-aineet
Ten to forty percent of the patients with acquired immunodeficiency syndrome (AIDS) develop cytomegalovirus (CMV) infections. In some patients with AIDS, CMV is detected in the bronchoalveolar lavage fluid (BALF), urine, and other specimens, even when there are no symptoms of CMV disease. An indicator of active CMV infection is needed to facilitate the diagnosis of CMV disease in patients with AIDS or HIV infection. CMV p65 antigen was detected in the leukocytes of both the peripheral blood and BALF during the early phase of CMV disease.

Type: Primary
Antigen: Not Applicable
Clonality:More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR