Anti-CD66 molecule Mouse Monoclonal Antibody (CF568) [clone: C66/1009]
Toimittaja: Biotium
BNC681009-500 BNC681009-100
BNC681009-500EA 560 EUR
BNC681009-500 BNC681009-100
Anti-CD66 molecule Mouse Monoclonal Antibody (CF568) [clone: C66/1009]
Vasta-aineet
This antibody recognizes proteins of 80-200 kDa, identified as different members of CEA family. CEA is synthesized during development in the fetal gut and is re-expressed in increased amounts in intestinal carcinomas and several other tumors. This MAb does not react with nonspecific cross-reacting antigen (NCA) and with human polymorphonuclear leucocytes. It shows no reaction with a variety of normal tissues and is suitable for staining of formalin/paraffin tissues. CEA is not found in benign glands, stroma, or malignant prostatic cells. Antibody to CEA is useful in detecting early foci ofMore Product Information