Anti-Double Stranded DNA Mouse Monoclonal Antibody (CF568) [clone: DSD/958]
Toimittaja: Biotium
BNC680958-500 BNC680958-100
BNC680958-500EA 560 EUR
BNC680958-500 BNC680958-100
Anti-Double Stranded DNA Mouse Monoclonal Antibody (CF568) [clone: DSD/958]
Vasta-aineet
This MAb recognizes the double stranded DNA in human cells. It can be used to stain the nuclei in cell or tissue preparations and can be used as a nuclear marker in human cells. This MAb produces a homogeneous staining pattern in the nucleus of normal and malignant cells. Deoxyribonucleic acid (DNA) is a nucleic acid that stores long-term information regarding the development and function of all known living organisms. DNA consists of two long nucleotide polymers, which are composed of four bases, namely adenine, thymine, guanine and cytosine, all of which are flanked by a phosphate-More Product Information