Anti-KRT7 Mouse Monoclonal Antibody (CF568) [clone: KRT7/903]

Toimittaja: Biotium
BNC680903-500 BNC680903-100
BNC680903-500EA 560 EUR
BNC680903-500 BNC680903-100
Anti-KRT7 Mouse Monoclonal Antibody (CF568) [clone: KRT7/903]
Vasta-aineet
Cytokeratin 7 is a basic cytokeratin, which is found in most glandular and transitional epithelia but not in the stratified squamous epithelia. Keratin 7 is expressed in the epithelial cells of ovary, lung, and breast but not of colon, prostate, or gastrointestinal tract. Antibody to cytokeratin is useful in distinguishing ovarian carcinomas (keratin 7 ) from colon carcinomas (keratin 7-).

Type: Primary
Antigen: KRT7
Clonality: Monoclonal
Clone: KRT7/903
Conjugation: CF568
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR