Anti-KRT19 Mouse Monoclonal Antibody (CF568) [clone: BA17]
Toimittaja: Biotium
BNC680666-500 BNC680666-100
BNC680666-500EA 560 EUR
BNC680666-500 BNC680666-100
Anti-KRT19 Mouse Monoclonal Antibody (CF568) [clone: BA17]
Vasta-aineet
This Ab reacts with the rod domain of human cytokeratin 19 (CK19), a polypeptide of 40 kDa. CK19 is expressed in sweat gland, mammary gland ductal and secretory cells, bile ducts, gastrointestinal tract, bladder urothelium, oral epithelia, esophagus, and ectocervical epithelium. Anti-CK19 reacts with a wide variety of epithelial malignancies including adenocarcinomas of the colon, stomach, pancreas, biliary tract, liver, and breast. Perhaps the most useful application is the identification of thyroid carcinoma of the papillary type, although 50%-60% of follicular carcinomas are also labeled.More Product Information