Anti-KRT14 Mouse Monoclonal Antibody (CF568) [clone: LL002]
Toimittaja: Biotium
BNC680499-500 BNC680499-100
BNC680499-500EA 560 EUR
BNC680499-500 BNC680499-100
Anti-KRT14 Mouse Monoclonal Antibody (CF568) [clone: LL002]
Vasta-aineet
Cytokeratin 14 (CK14) belongs to the type I (or A or acidic) subfamily of low molecular weight keratins and exists in combination with keratin 5 (type II or B or basic). CK14 is found in basal cells of squamous epithelia, some glandular epithelia, myoepithelium, and mesothelial cells. Anti-CK14 is useful in differentiating squamous cell carcinomas from poorly differentiated epithelial tumors. Anti-CK14 is one of the specific basal markers for distinguishing between basal and non-basal subtypes of breast carcinomas. Anti-CK14 is also a good marker for differentiation of intraductal fromMore Product Information